Biocam Virus Chart Set
Supplier: BioCam Communications
SPECIAL
470023-544KT 115 USD
470023-544
Biocam Virus Chart Set
Charts
Comprehensive Overview of Structure and Replication
  • Colorful illustrations and descriptive text
These charts present useful information for your virology studies. The Virus I chart depicts well-known examples of icosahedral, helical, enveloped, and complex virus structures; Virus II chart covers replication, with detail on Influenza life cycle; Virus III presents information specific to bacteriophages, contrasting lytic and lysogenic cycles. Set of three charts.