Dressing Stick for Grinders
Supplier: SAINT GOBAIN ABRASIVES
61463610599
470022-256EA 49.65 USD
470022-256
Dressing Stick for Grinders
Scissors
Extends Life of Diamond Cup Wheel.
A silicon carbide stick helps keep your grinder working smoothly. Size: 1/2 ×1/2×6".