Dressing Stick for Cutoff Saws
Supplier: SAINT GOBAIN ABRASIVES
281927
470022-254EA 35.95 USD
470022-254
Dressing Stick for Cutoff Saws
Cutters
Keeps diamond blades free of buildup.
Blades last longer and cut faster with this specially bonded silicon carbide.