Coolant Pump System
Supplier: GENERAL MACHINE/DIECRON, INC.
28-1920
470022-250EA 1508.3 USD
470022-250
Coolant Pump System
Pumps Drum/Transfer Pumps
Self-Contained Unit for Use with Any Saw/Grinder
Make your thin section processing lab more mobile with this unit that requires no water hookups for cooling. The easy-to-maintain, five-gallon reservoir tank is removable for cleaning, and the cutoff valve prevents spilling when disconnected from pump. The quiet, dependable motor pumps 325 gallons per hour at 1' rise. Required for all thin section saws/grinders sold by WARD’S. Size: 19"L x 19"W x 15"H. 120 VAC, 60 Hz. Weight: 33 lbs.