Eyepiece Pointer Wire
Supplier: Walter Products
PT101
470021-924EA 14.5 USD
470021-924
Eyepiece Pointer Wire
Microscope Eyepieces
The pointer is housed in a disc that fits into your microscope's eyepiece, and is held in place by a spring. Point out specific structures under the microscope.