Magni-Grip Magnifier with Forceps
Supplier: CARSON OPTICAL
MG-55
470021-818EA 9.4 USD
470021-818
Magni-Grip Magnifier with Forceps
Magnifiers
Perform Fine Dissections and Handle Small Specimens Easily
The 4X magnifying lens is attached to fine-tipped stainless steel forceps to view even very small specimens. The forceps act as a base for the magnifier for hands-free observation. It folds for storage. Size: 4 1/4"L x 1 1/2"W, lens size 1 1/8" dia., forceps size 3 1/4"L.