Peak Loupe
Supplier: Selsi
1962
470021-810EA 123.05 USD
470021-810
Peak Loupe
Magnifiers
Large Lenses for Wide, Distortion-Free Viewing Area
The clear plastic base of this convenient loupe can be placed over a live specimen to contain it for closer examination. Size: 1 1/4"L x 1 3/8" widest diameter. 15X magnification.