LaMotte® Soil pH TesTabs Kit
Supplier: LA MOTTE CHEMICAL PROD CO SE
5912
470021-532EA 22.4 USD
470021-532
LaMotte® Soil pH TesTabs Kit
Standards Environmental Analysis Standards
An Ideal Alternative to Liquid pH Indicators
  • No hazardous compounds required for testing
  • Colorimetric pH determination
  • Includes instructions
Quickly and simply measure soil pH with these non-hazardous reagents. The kit includes 50 tests and a laminated color chart for comparisons and pH determinations. Chart values feature range 4.0 to 11.0 pH. Distilled or deionized water recommended. Includes instructions.