Ammonia Test Kit
Supplier: Pet World Inc
LR8600
470021-372KT 19.25 USD
470021-372
Ammonia Test Kit
Specimens Live Specimens Aquaria - Living Material
Fast and easy-to-use, this kit contains two dropper bottles for accurate dispensing of test solutions, non-staining test tube with snap cap, and instructions. It comes with enough solution for 130 tests and can be used in either fresh or salt water. Range: 0–8 ppm ammonia.