Zebra Fish Breeding Tank
Supplier: AQUANEERING LLC
ZHCT100
470021-368EA 59.05 USD
470021-368
Zebra Fish Breeding Tank
Tanks Aquarium Tanks
Great For Overnight Breeding Of Zebra Fish
The breeding setup consists of one flexible tank with a screened bottom nesting inside a second flexible tank and covered with a lid. The inner tank is screened to allow the eggs to fall into the bottom tank so the adults will not eat them.