Fieldmaster Mini-Surber Bottom Sampling Net
Supplier: Science First
623-1210
470021-242EA 166 USD
470021-242
Fieldmaster Mini-Surber Bottom Sampling Net
Samplers Water Samplers
Smaller Size for Benthic Invertebrates
  • Quantitative Sampling
One of the best quantitative samplers, this net is normally used for macroinvertebrates and sometines larger specimens like crayfish, adult insects, and more. Also called a riffle sampler or run sampler, the Mini-Surber is designed for small streams and rivulets where larger models will not fit. Constructed of genuine Nitex bolting cloth with taffeta wings with a stainless steel frame, the net provides maximum corrosion resistance. Size: 10 1/2" long.