Tree Tape
Supplier: Peco Sales
59471
470021-028EA 81.35 USD
470021-028
Tree Tape
Educational Materials Biology Educational Materials Ecology Learning Activities
Determine the Diameter of Trunks by Measuring their Circumference
This chrome-plated steel tape comes with a claw hook for fastening the tape to trees. Graduations are marked in 10ths, 100ths, and whole feet, while the reverse side of the tape gives diameter equivalents up to 120". The tape can be rewound into its heavy-duty, plastic case with the handle, which fits neatly into the case when not in use. The claw hook also fits into the side of the case when it is not in use. Tape size: 35'L x 3/8"W; case size: 3 1/2" dia. x 3/4".