Foam Plugs
Supplier: Avantor
470020-896PK 9.5 USD
470020-896
Foam Plugs
Stoppers Cellulose Stoppers
These foam plugs are designed to fit most test tubes and vials.
They are ideal for culture applications and can be sterilized and reused.