Polypropylene Petri Dish Rack
Supplier: Bel-Art
F18991-0000_SE
470020-886EA 129.6 USD
470020-886
Polypropylene Petri Dish Rack
Culture Dishes Petri Dishes
This autoclavable polypropylene rack holds forty-two 100 mm dishes. It has a small footprint, is stackable to conserve counter space, and it features a handle for easy transport. Size: 13"L x 8 1/4"W x 7"H.