Polyhedral Alcohol Burner
Supplier: American Scientific
2066-11
470020-698EA 18 USD
470020-698
Polyhedral Alcohol Burner
Heaters Spirit Lamps
Position the Flame at Just the Right Angle.
  • Capacity: 125 mL
  • Height: 4"
The polyhedral shape of this heavy-duty glass burner allows you to position the flame at just the right angle for different tasks. It comes with a nickel-plated snuffer cap and one wick. Uses 1/4" (6mm) thick replacement wicks.