Slotted Microscope Slide Staining Dish
Supplier: C & A Scientific
SD-10
470020-694EA 13.65 USD
470020-694
Slotted Microscope Slide Staining Dish
Jars Staining Jars
Durable Staining Dish with Less Mess
  • Quality slide making supplies
  • Slide staining tools
This deep covered glass dish lessens the risk of overflow. It will hold 20 slides back to back or ten single slides.