Lampboard
Supplier: United Scientific Supplies
LMPB05 LMPB03
470020-576EA 23.35 USD
470020-576 470014-932
Lampboard
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Study series and parallel circuits with this high quality apparatus.
  • Includes lamps and an activity manual.
  • 3 bulbs included
  • Recommended bulb size is 1.5V and 3.8V (0.3 amps)
The connecting strips allow students to switch between series and parallel circuits. Comes on an acrylic base. Measures 5" x 10"