Ohaus Starter Pen Salinity Meters
Supplier: Ohaus
30073982 30073983 30073982 30073983
470020-292EA 106.5 USD
470020-292 470020-302 77681-996 77681-998
Ohaus Starter Pen Salinity Meters
Electrochemistry Meters Salinity Meters
When you are looking for accurate results anywhere and anytime, look no further than the OHAUS Starter Series of pen meters designed for education.
  • Wrist Strap Prevents Unintentional Dropping
  • Waterproof Design
  • Protective Cap
They're simple, fast, and straightforward.
With durable plastic housing, a protective cap that safeguards the sensor, and an automatic shutdown feature to preserve battery life, the Starter pen meters can endure consistent use in rugged environments.