Latent Fingerprinting Supplies
Supplier: FORENSICS SOURCE
1-0300 0196-00001 0187-00001
470020-198EA 7.15 USD
470020-198 470149-780 470149-656
Latent Fingerprinting Supplies
Educational Materials Forensics Educational Materials Forensics Supplies
Develop and study otherwise scarcely visible fingerprints using high-quality fingerprint brushes.
  • Different fingerprinting brushes to suit your preferences
  • Supplies Demonstrate the minute differences in individual fingerprints
  • Camel Brush Ideal for General Conditions
  • Zephyr Brush Retains a Large Amount of Powder
  • Brush Length: 61/4"
For general conditions, the camel hair brush is ideal.
The Zephyr fiberglass brush retains a larger amount of powder and applies it more evenly to provide clear prints in any circumstance.