Fast-Drying Epoxy Mounting Medium
Supplier: ELLSWORTH ADHESIVE
608
470020-092KT 42.29 USD
470020-092
Fast-Drying Epoxy Mounting Medium
Stains Microscopy and staining reagents
Process Slides in Just One Day
For fast results, mix the hardener (1.4 oz.) and resin (1.4 oz.), mount the chip to the slide, and the section will set in five minutes! Curing takes just 1–2 hours.