Epoxy Cement
Supplier: ELLSWORTH ADHESIVE
83176
470020-090EA 52.85 USD
470020-090
Epoxy Cement
Adhesives
This is an epoxy cement kit.
  • Kit includes both resin and hardener tubes
  • Resin: 2 ½ oz.
  • Hardener: ¾ oz.
The epoxy will cure in a day, or in just hours at higher temperatures.