Disposable Micropipette Tips
Supplier: Fischer Technical
D592 637-2
470019-832EA 94.15 USD
470019-832 470233-654
Disposable Micropipette Tips
Pipette Tips
High quality micropipette tips to meet any lab's needs.
  • Certified DNase, RNase and pyrogen free
  • Material: PP
  • Autoclavable
  • Designed to fit most standard micropipettes
All quantities are supplied in bulk bags.