Lab Label Roll
Supplier: STAPLES CONTRACT&COMMERCIALINC
130575 15-1970
470019-780EA 5 USD
470019-780 470019-788
Lab Label Roll
Labels
Better than gummed labels, these pressure-sensitive labels adhere securely, yet can be easily removed.
  • Autoclavable to 260 °C
The labels are on a continuous roll.