Portable Waterproof pH/EC/TDS Meter, Low Range
Supplier: HANNA INSTRUMENTS
HI991300
470019-690EA 567.19 USD
470019-690
Portable Waterproof pH/EC/TDS Meter, Low Range
Electrochemistry Meters pH and Multi-Parameter Meters
Designed to bring simplicity to environmental testing, this pH/EC/TDS Meter is designed specifically for use in manufacturing and environmental testing protocols. The specialized electrode combines 3 sensors in a single rugged probe body featuring a polypropylene body, integrated temperature sensor, and cloth junction.
  • 3-in-1 probe measures pH, EC/TDS, and temperature
  • Comes with all the necessary solutions and batteries - everything you need to get started measuring right away
  • Perfect for water quality testing
  • Probe condition - an on-screen indicator provides visual confirmation that your probe is working at its best
This meter is designed to withstand the knocks, drops, and spills of real life, the new IP 67 body ensures top performance in any environment. These meters are protected against dust and water intrusion from any direction. Also has a portable design, Made to fit comfortably in your hand for on-the-go testing, the multilevel LCD display provides at-a-glance readings of pH and temperature from any angle and dual button operation streamlines the testing process for any user. The probe connection is a quick Din connection. A Quick Connect DIN connector makes attaching and removing the probe simple and easy. The rubber coating protects the cable and provides a water-tight seal without twisting.
This pH/EC/TDS meter features a replaceable HI12883 probe that has been carefully designed specifically for pH measurements encountered in manufacturing and environmental testing protocols. The polypropylene body of the HI12883 houses all three sensors within a sturdy, single body design. The plastic body protects the electrode from damage, is easy to clean, and is chemically resistant to many aggressive chemicals. The pH sensor in the probe is a single junction electrode with gelled electrolyte and a retractable cloth junction. When readings appear sluggish, simply pull out some of the cloth material from out from its position t renew the junction with an uncontaminated, fresh surface. The EC/TDS portion is a two-ring probe that comes with the HI991300, offering an accurate solution for low range EC and TDS readings. An integrated temperature probe ensures fast, accurately compensated readings even during sudden temperature fluctuations.
Low battery indicator - the included batteries provide 1400 hours of continuous use. When the battery level is below 10%, a warning symbol will blink to indicate low battery condition. When low battery power may affect your results, the meter will shut down to avoid erroneous readings caused by low battery level.
Ordering information: Portable pH/EC/TDS meter comes with cleaning solution that has been engineered to keep your electrode free from debris and build up. This concentrated combination of detergents cleans your meter quickly and effectively so that your meter stays in optimum condition. When performing a pH calibration it is important that the correct buffers are used. We include some start buffers to get you going. The single-use sachets are individually sealed so you use fresh buffer each time you calibrate. When performing an EC/TDS calibration it is important that the correct standards are used. We include some starter standards to get you going. The single-use sachets are individually sealed so you use fresh buffer each time you calibrate.