Mini Stirrer
Supplier: HANNA INSTRUMENTS
HI200M-1
470019-666EA 95.79 USD
470019-666
Mini Stirrer
Stirrers Magnetic Hotplate Stirrers
Control stirring speed with precision.
  • Both plastic and steel surfaces are chemically resistant
  • Speed limiter for safety with max speed of 1000 rpm
  • 1 L capacity