Lab Burner
Supplier: Wards
CH2019001
470019-572EA 43.85 USD
470019-572
Lab Burner
Heaters Gas Burners
These laboratory burners are compact with a powerful and concentrated flame.
  • Made of brass and stainless steel
Soot-free use and adjustable flame control. The burners screw into a cartridge to guarantee a leak free installation.
Burner has separate adjustments for regulating gas and air intake. Flame top: 1¼" diameter, burner unit: 2¼"×2⅔".