Fahrenheit Red Alcohol Thermometers
Supplier: Thermco
CSP2040P2S
470019-556BX 169 USD
470019-556
Fahrenheit Red Alcohol Thermometers
Thermometers Analog Thermometers
Partial immersion thermometer with pocket case.
Range: -40° to +120°F
Graduations: 2°F