Tripod Ring Stands
Supplier: United Scientific Supplies
TRS4X9
470019-528EA 26.05 USD
470019-528
Tripod Ring Stands
Tripods
Support stand for holding labware over a burner.
  • Cast iron ring
  • Detachable legs
  • Zinc plated legs
Hold beakers and flasks at a safe distance from the heat source such as a bunsen burner using these tripods. Each comes standard with one ring and detachable legs. Size: 9"H.