Ward's® Osteometric Board
Supplier: Dynalon
8551-3224
470019-330EA 314 USD
470019-330
Ward's® Osteometric Board
Models Human Biology Models
Use a Tool of Forensic Scientists at an Economical Price
  • Easy to Transport
  • Affordable
Made of durable, lightweight polypropylene, this osteometric board is the most affordable on the market.
The sturdy fixed headpiece and sliding footpiece are operated with a smooth, fluid motion to allow for precise and accurate measurement of long bones up to 60cm(600mm), and it features an accurate, easy-to-read scale in mm to eliminate the need for conversions in standard regression formulas.
Size: 30"L x 9"W x 4"H