InstaPulse® Heart Rate Monitor
Supplier: BIOSIG INSTRUMENTS
105
470019-162EA 204.25 USD
470019-162
InstaPulse® Heart Rate Monitor
Educational Materials Biology Educational Materials Physiology Learning Activities
Instant Readings Give The First Readout After Four Heartbeats
This monitor can be used any time, even during exercise and is an excellent tool for studying trends, irregularities, or even learning biofeedback. It is so easy to use that all you need to do is grasp the rings and it turns on. When you let go, it turns off. The crystal controlled accuracy never needs calibration and features a range of 30–240 bpm. It comes in a water-tight casing and includes a 9 V alkaline (transistor) battery and instructions.