Hydraulic Hand Dynamometer, FABRICATION ENTERPRISES INC.
Supplier: Fabrication Enterprises
12-0240
470019-094CS 3751.7 USD
470019-094
Hydraulic Hand Dynamometer, FABRICATION ENTERPRISES INC.
Dynamometers
Used By Professional Physical Therapists for Reliability and Repeatability
Receive accurate grip strength measurements without feeling the handle move, with the hydraulic and bellows operation of this instrument. A five position adjustable handle accommodates any hand size, while a large, dual-scale gauge features a maximum reading indicator. Range: 0-200 lbs in 2 lb increments; 0-90 kg in 5 kg increments. The all metal construction provides durability. Comes in a protective carrying case with manual. Size: 10"L x 2 1/2" dia.