Instant Ear Thermometer
Supplier: Graham-Field
2215
470018-918EA 54.8 USD
470018-918
Instant Ear Thermometer
Thermometers Digital Thermometers
This thermometer features one-touch operation and avoids the hassle of oral thermometers.
  • Accurate Readout in One Second
  • Fever Alarm
The user-friendly instrument yields accurate results within one second of being placed in the patient’s ear. A fever alarm sounds for temperatures above 100.4°F. The thermometer's display indicates measurements in either Celsius or Fahrenheit, and the Nite-Glo display can be seen in any lighting. The thermometer has a range of 93.2°F to 109.4°F and includes a storage case, lithium battery, and guidebook.