Dissecting Needles
Supplier: Lab Connections
B235-2 WLS26805 NEDPL1-PK/12
470018-900EA 2 USD
470018-900 470147-516 470152-804
Dissecting Needles
Dissection Needles
A must for any dissection.
  • Use to tease muscle or tissue apart
  • Selection of wood, metal, or plastic handles
  • Straight or bent tips
Choose a size to suit your personal preference. Straight needles are used for general work, while bent tips make tracing vessels and nerves easy. Available in all stainless steel, or stainless needles in plastic or wood handles.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.