Slim Taper Triangular Files with Wood Handle
Supplier: United Scientific Supplies
TRF004
470018-892EA 6.45 USD
470018-892 470018-896 470200-926 470201-366
Slim Taper Triangular Files with Wood Handle
Glass Files
Useful for metal filing or glass cutting.
  • Wooden handle for easy gripping
  • Hardened triple sides
Triangular file has a tapered shape, and is mounted on a hardwood handle for easy gripping. For general laboratory use, including to score glass before cutting.