Insect Pinning Forceps
Supplier: Dunrite Instruments
140250
470018-852EA 6.1 USD
470018-852
Insect Pinning Forceps
Forceps
Designed To Handle And Pin Insects
The broad, angled tips of these forceps are ideal for inserting between wing of folded specimens, to permit easy and damage-free pinning and mounting. Arkhurst Style with very broad serrated bent tips.