Aluminum Slide Holders
Supplier: HUGH COURTRIGHT & CO.
28-A
470018-786PK 165.85 USD
470018-786 470018-788
Aluminum Slide Holders
Slide Holders
Converts cardboard micropaleontology slides into permanent displays.
These slide holders secure a 1" x 3" glass slide to cardboard for protection. Packages of 100.