Fieldmaster Metal Sieve Set
Supplier: Science First
621-7110
470018-758KT 239.35 USD
470018-758
Fieldmaster Metal Sieve Set
Sieves Test Sieves
Stacking Set for Simple Storage
These economical stainless steel mesh sieves can be stacked together to help separate soil or sediment particle sizes. The set of six includes the following sizes: 4000 micron, 2000 micron, 500 micron, 250 micron, 125 micron and 63 micron. A lid and a catch pan are also included.