Electric Sieve Shaker
Supplier: Humboldt Manufacturing
H-4325
470018-706EA 2170.95 USD
470018-706
Electric Sieve Shaker
Shakers and Mixers Sieve Shakers
Assure Accurate and Dependable Separations Every Time
Our electric sieve shaker is specially designed for both vertical and horizontal tapping action. Spring-loaded tie rods and a 30-minute, built-in timer offer a steady, regulated shaking action. This unit has a larger capacity than most models, accommodating up to nine standard 8" sieves, not included, as well as a bottom pan and lid. A built-in adapter and convenient lid lock allow the unit to hold fewer sieves, or to hold smaller 3" or 6" diameter sieves. It is powered by a quiet-running, beltdriven, 1/4 hp motor. 115 VAC, with a 5 1/2' cord; 60 Hz. Size 14"L x 12"W x 36"H; weight: 65 lbs.