Brunton Com/Pro Lightweight Pocket Transits
Supplier: Brunton USA
5008 5007
470018-654EA 606.09 USD
470018-654 470018-656
Brunton Com/Pro Lightweight Pocket Transits
Compasses
Versatile Units Incorporate Several Tools in One.
  • Vertical angle measurements to ±90°
  • Induction dampened needle for quick, accurate readings
  • Precision aligned mirror with convenient "see-through" sighting capability
  • Magnetic declination adjustments allow for east or west declination settings to ±180°
Enjoy many of the features of a professional transit: Brunton transits with induction damping combine the principles of a surveyor’s compass, a prismatic compass, clinometer, hand level, and plumb. For additional precision, transits can be adjusted for magnetic declination with a simple screw turn on the case.
The dial is graduated in 1° increments to obtain closer readings than from standard 2 to 5° graduated models. For life-long utility, units include a lift-pin mechanism to protect parts from accidental impact; rubber O-rings keep out moisture and dirt. The sleek composite plastic housing is as durable as metal but lighter in weight.
Ordering information: Supplied with leather case and instruction.