Charger for Killing Jar
Supplier: Ward's Science
EE0068-KIT
470018-456PK 34.75 USD
470018-456
Charger for Killing Jar
Jars Specimen Jars
Includes Squeeze Bottle for Convenient Application
Ethyl acetate’s relatively low toxicity to humans, yet quick action on insects makes this a recommended killing agent. The package includes a refillable 1 oz. plastic squeeze bottle with 100 mL of ethyl acetate. With MSDS. Note: Shipping and drug restrictions apply. Sold on institutional purchase order only.