Universal Precaution Compliance Kit
Supplier: MASUNE INC.
19151M
470018-446KT 44.2 USD
470018-446
Universal Precaution Compliance Kit
Spill Control Materials
Protect yourself, and clean up your lab, with one handy combination kit. Items for protection against pathogen spills, as well as equipment for safe removal and disposal of spilled materials, all of which meet OSHA, CDC, and state requirements, are provided. The kit includes a safety shield with mask, disposable apron, medical gloves, Red Z pouch, scoop with scraper, germicidal disinfectant surface wipe, biohazard waste bag, antimicrobial hand wipe, and identification tag.