Essential Physics Demo: Venturi Tube
Supplier: VWR International
PH0137VD
470018-310EA 71.4 USD
470018-310
Essential Physics Demo: Venturi Tube
Educational Materials Physics Educational Materials Mechanics Learning Activities
Investigate the relationship between the speed of a fluid and its pressure
  • Easy to use
This impressive demonstration consists of a glass Ventri tube with two different diameters, with a set of additional tubes attached to the different sections . One pair holds a U-shaped tube in which a color liquid can be placed to demonstrate the different pressures exerted when air is blown through the tube.
Ordering information: Demo includes two rubber hoses, glass Venturi tube, U-shaped tube and two hose clamps.