Double-Walled Calorimeter
Supplier: PRECISION MOULDED POLYMERS LIMITED
1350401010000
470018-276EA 36.7 USD
470018-276
Double-Walled Calorimeter
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Simple Calorimeter to Measure Heat in Many Applications
The double-walled cover minimizes heat loss, and the inner and outer vessels are both made of aluminum. The inner vessel, which has a 200 mL capacity, comes with a stirrer and support.