Student Grade Kick Net
Supplier: LaMotte
0172
470017-956EA 54.7 USD
470017-956
Student Grade Kick Net
Animal Supplies Animal Nets
Easy Stream Sampling
  • Collect Samples From theStream Bottom
Ideal for student macroinvertebrate collection, this durable mesh net measures approximately 35" x 45". Three inch pockets are featured on opposite sides to hold poles in place. Poles are not supplied with unit, but can be purchased at any local hardware store. Mesh size is 1/32".