Vernier® Thermocouple
Supplier: Vernier Software & Technology
TCA-BTA
470017-896EA 78.99 USD
470017-896
Vernier® Thermocouple
Temperature Probes
This sensor uses type-K thermocouple wire to measure temperatures over the range of −200 to 1400 °C.
It can be used to measure flame temperatures as high as 1400°C, or liquid nitrogen temperatures at -196°C. An internal ice-point compensation chip is included, so you do not need to place a reference wire in an ice-water bath. You can simply use one measuring lead to take temperature readings. Each thermocouple is individually calibrated.