Vernier® ORP Sensor
Supplier: Vernier Software & Technology
ORP-BTA
470017-890EA 114.99 USD
470017-890
Vernier® ORP Sensor
Electrodes Redox Electrodes
Measure the ability of a solution to act as an oxidizing agent or reducing agent with this oxidation-reduction potential sensor.
Positive readings indicate an oxidizing agent, while negative readings indicate a reducing agent. ORP electrodes are often used to measure the oxidizing ability of chlorine in swimming pools, or to determine when the equivalence point has been reached in an oxidation-reduction reaction.
Range: -450 to +1100 mV.