Purple Starchy:Purple Sweet:Yellow Starchy:Yelllow Sweet 1:1:1:1 Ratio
Supplier: Ward's Science
WF-17-6602
470017-836EA 23.8 USD
470017-836
Purple Starchy:Purple Sweet:Yellow Starchy:Yelllow Sweet 1:1:1:1 Ratio
Specimens Live Specimens Plants - Living Material
Purple Starchy: Purple Sweet: Yellow Starchy: Yelllow Sweet. The result of test crossing the F1 hybrid with the recessive parent (R/r Su/su and r/r su/su).
  • Genetics
Visual Example of Genetics at Work
Dried and shrink-wrapped ear of corn illustrates naturally occurring phenotype ratios of genetic crosses.