Filter Support Collar
Supplier: United Scientific Supplies
FLTR01
470017-722EA 12.35 USD
470017-722
Filter Support Collar
Supports Support Sleeves
Filter Support Collar is a sturdy, molded rubber filter disc.
  • Inside Diameter: 32 mm
  • Outside Diameter: 76 mm
  • Molded Rubber Construction
It is designed to support and seal a filter crucible or funnel in the neck of a filter flask.