Molecular Orbital Models
Supplier: United Scientific Supplies
MOMS07
470017-652EA 179.75 USD
470017-652
Molecular Orbital Models
Models Molecular Biology Models
The most common atomic and hybridized molecular orbitals are illustrated in this set of seven models.
  • Models Mounted on Z-axis
  • Removable
  • Seven Models per Set
Each model, which is mounted on a removable stand, is assembled on a vertical rod that represents the Z-axis.
Models include the following orbitals: S, PX, PY, PZ, SP1, SP2, and SP3.