Vernier® Cart Picket Fence
Supplier: Vernier Software & Technology
PF-CART
470017-616EA 5.99 USD
470017-616
Vernier® Cart Picket Fence
Educational Materials Physics Educational Materials Mechanics Learning Activities
Accurately measure the velocity of your dynamics cart.
  • Dual marking scales: one 50 mm bar, and 13 alternating clear and black bars, with an interval of 10 mm
  • Intended for use with Vernier dynamics carts
The cart picket fence is a small plastic strip for doing photogate motion timing with dynamics carts. When the cart picket fence passes through a photogate, the cart’s velocity and acceleration as a function of time can be accurately measured.
Ordering information: Vernier dynamics cart and photogates available separately.